• USA Home
  • W269700 - α-Methylcinnamaldehyde

EMAIL THIS PAGE TO A FRIEND
W269700 Aldrich

α-Methylcinnamaldehyde

≥97%, FCC, FG

Synonym: α-Methylcinnamaldehyde

  • CAS Number 101-39-3

  • Empirical Formula (Hill Notation) C10H10O

  • Molecular Weight 146.19

  •  Council of Europe no. 578

  •  EC Number 202-938-8

  •  FEMA Number 2697

  •  Flavis number 5.050

  •  PubChem Substance ID 24901264

  • Popular Documents:  Specification Sheet (PDF)  

Purchase

Properties

Related Categories A-D, Alphabetical Listings, Certified Food Grade Products, Flavors and Fragrances, Kosher Certified Products,
biological source   synthetic
grade   FG
  Kosher
reg. compliance   FCC
  FDA 21 CFR (172.515)
  EU Regulation 1334/2008 & 872/2012
InChI Key   VLUMOWNVWOXZAU-VQHVLOKHSA-N
assay   ≥97%
maycontain   ≤0.050% a-tocopherol as antioxidant
refractive index   n20/D 1.6045
bp   148-149 °C/27 mmHg
density   1.037 g/mL at 25 °C
GMO content   GMO-free
Organoleptic   cinnamon; spicy; herbaceous; sweet
food allergen   no known allergens

Description

Packaging

1 kg in glass bottle

10 kg in poly drum

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
GD6600000
Flash Point(F) 
174.2 °F
Flash Point(C) 
79 °C
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?