EMAIL THIS PAGE TO A FRIEND
W292435 Aldrich

Propionic acid

99%, FCC

Synonym: Acid C3, Propanoic acid, Propanyl acid

  • CAS Number 79-09-4

  • Linear Formula CH3CH2COOH

  • Molecular Weight 74.08

  •  Beilstein Registry Number 506071

  •  Council of Europe no. 3c

  •  EC Number 201-176-3

  •  FEMA Number 2924

  •  Flavis number 8.003

  •  MDL number MFCD00002756

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, O-P More...
biological source   synthetic
reg. compliance   FCC
vapor density   2.55 (vs air)
vapor pressure   2.4 mmHg ( 20 °C)
InChI Key   XBDQKXXYIPTUBI-UHFFFAOYSA-N
assay   99%
autoignition temp.   955 °F
expl. lim.   12.1 %
refractive index   n20/D 1.386(lit.)
  n20/D 1.386
bp   141 °C(lit.)
  141 °C
mp   −24-−23 °C(lit.)
density   0.993 g/mL at 25 °C(lit.)
  0.995 g/mL at 25 °C

Description

Legal Information

Manufactured by Eastman Chemical Company.

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3463 8/PG 2
WGK Germany 
1
RTECS 
UE5950000
Flash Point(F) 
129.2 °F
Flash Point(C) 
54 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Food Safety

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

W292443 Propionic acid, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?