• USA Home
  • W322407 - 2-Phenyl-2-butenal, mixture of cis and trans

EMAIL THIS PAGE TO A FRIEND
W322407 Aldrich

2-Phenyl-2-butenal, mixture of cis and trans

≥97%, FG

Purchase

Properties

Related Categories Alphabetical Listings, Certified Food Grade Products, Flavors and Fragrances, Halal Certified Products, N-P,
biological source   synthetic
grade   FG
  Halal
reg. compliance   FDA 21 CFR (110)
  EU Regulation 1334/2008 & 872/2012
InChI Key   DYAOGZLLMZQVHY-UHFFFAOYSA-N
assay   ≥97%
refractive index   n20/D 1.561(lit.)
bp   177 °C/15 mmHg(lit.)
density   1.034 g/mL at 25 °C(lit.)
Organoleptic   chocolate; earthy; floral

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
222.8 °F
Flash Point(C) 
106 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Food Safety

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?