• USA Home
  • W328915 - cis-4-Heptenal solution

EMAIL THIS PAGE TO A FRIEND
W328915 Aldrich

cis-4-Heptenal solution

10 wt. % in triethyl citrate, natural (US), FG

Purchase

Properties

Related Categories A-D, Alphabetical Listings, C-D, Certified Food Grade Products, Certified Natural Products,
biological source   synthetic
grade   FG
  Kosher
  natural (US)
reg. compliance   FDA 21 CFR (172.515)
  EU Regulation 1334/2008 & 178/2002
InChI Key   VVGOCOMZRGWHPI-ARJAWSKDSA-N
concentration   10 wt. % in triethyl citrate
density   1.080 g/mL at 25 °C
Certification   Kosher, GMO-free, food allergen
See Safety & Documentation for available documents
Organoleptic   fatty; green; vegetable; potato
food allergen   no known allergens

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability


New Product Offerings
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Potential Application for CIS-4 Heptenal, 10 WT.% in Triethyl Citrate, FG

Use this flavor additive mainly in dairy products where it lends top notes to sour creams, cultured butters, vanilla creams and ghee flavors. It also finds applications in potato, olive oil and spear...

Securing a Traceable Supply Chain for Food Grade Flavor Ingredients

Introduction Defining "Food Grade" Understanding "Food Grade" Versus "Industrial Grade" Top Questions to Ask a Flavor & Fragrances Supplier The Sigma-Aldrich Approach
Dr. Luke Grocholl
Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Food & Beverage, Food Safety, Solvents

The Complex Regulatory Landscape for Natural Flavor Ingredients

Introduction The Regulatory Landscape US and EU Definitions of Natural Regional Regulatory Variations The Example of Vanillin The Case for Standardization
Mr. Michael Sabisch1, Mr. David Smith2

1Senior Quality Assurance Specialist
2International Sales Coordinator, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food Safety, Oxidations

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?