• USA Home
  • W367702 - Ethyl 3-mercaptopropionate

EMAIL THIS PAGE TO A FRIEND
W367702 Aldrich

Ethyl 3-mercaptopropionate

≥99%

  • CAS Number 5466-06-8

  • Linear Formula HSCH2CH2CO2C2H5

  • Molecular Weight 134.20

  •  EC Number 226-771-5

  •  FEMA Number 3677

  •  Flavis number 12.083

  •  MDL number MFCD00004896

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Alphabetical Listings, E-F, Flavors and Fragrances More...
biological source   synthetic
InChI Key   CJQWLNNCQIHKHP-UHFFFAOYSA-N
assay   ≥99%
refractive index   n20/D 1.457(lit.)
bp   75-76 °C/10 mmHg(lit.)
density   1.059 g/mL at 20 °C(lit.)

Description

Features and Benefits

Skunk, meaty

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
163 °F
Flash Point(C) 
73 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Food Safety

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?