• USA Home
  • W502308 - 2-Acetyl-3-methylpyrazine

EMAIL THIS PAGE TO A FRIEND
W502308 Aldrich

2-Acetyl-3-methylpyrazine

≥98%

Purchase

Properties

biological source   synthetic
InChI Key   QUNOTZOHYZZWKQ-UHFFFAOYSA-N
assay   ≥98%
refractive index   n20/D 1.521(lit.)
bp   90 °C/20 mmHg(lit.)
density   1.114 g/mL at 25 °C(lit.)
Organoleptic   vegetable; nutty

Description

Features and Benefits

Roasted, nutty, vegetable, toasted grain

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
176 °F
Flash Point(C) 
80 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?