• USA Home
  • W507997 - 2′-Aminoacetophenone

EMAIL THIS PAGE TO A FRIEND
W507997 Aldrich

2′-Aminoacetophenone

≥98%

Synonym: 2-Acetylaniline

  • CAS Number 551-93-9

  • Linear Formula H2NC6H4COCH3

  • Molecular Weight 135.16

  •  Beilstein Registry Number 386122

  •  EC Number 209-002-8

  •  FEMA Number 3906

  •  Flavis number 11.008

  •  MDL number MFCD00007717

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories A-B, Alphabetical Listings, Flavors and Fragrances More...
biological source   synthetic
InChI Key   GTDQGKWDWVUKTI-UHFFFAOYSA-N
assay   ≥98%
refractive index   n20/D 1.614(lit.)
bp   85-90 °C/0.5 mmHg(lit.)
density   1.112 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
235 °F
Flash Point(C) 
113 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Food Safety

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?