EMAIL THIS PAGE TO A FRIEND
W508918 Aldrich

Pinacol

≥98%

Synonym: 2,3-Dimethyl-2,3-butanediol, Tetramethylethylene glycol

  • CAS Number 76-09-5

  • Linear Formula HOC(CH3)2C(CH3)2OH

  • Molecular Weight 118.17

  •  Beilstein Registry Number 1340501

  •  EC Number 200-933-5

  •  MDL number MFCD00004462

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, O-P More...
biological source   synthetic
InChI Key   IVDFJHOHABJVEH-UHFFFAOYSA-N
assay   ≥98%
bp   171-172 °C/739 mmHg(lit.)
mp   40-43 °C(lit.)

Description

Features and Benefits

Sweet, warm bread, aromatic

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN1325 - class 4.1 - PG 2 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
2
RTECS 
EK1720000
Flash Point(F) 
170.6 °F
Flash Point(C) 
77 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?