EMAIL THIS PAGE TO A FRIEND
W510483 Aldrich

Sassafras oil

Purchase

Properties

Related Categories Alphabetical Listings, Essential Oils, Flavors and Fragrances, Halal Certified Products, Kosher Certified Products,
biological source   from Sassafras albidum (Nutt) Nees
grade   Halal
  Kosher
description   contains >90% Safrole
origin   Vietnam origin
refractive index   n20/D 1.536(lit.)
bp   236 °C(lit.)
density   1.09 g/mL at 25 °C(lit.)
GMO content   GMO-free
Organoleptic   spicy
food allergen   no known allergens

Description

Application

Not intended for use as a food flavoring additive.

Preparation Note

Extraction method: steam distillation of Sassafras tree roots.

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Food Safety

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?