EMAIL THIS PAGE TO A FRIEND
W520306 Aldrich

Hexyl salicylate

>98%

Synonym: Hexyl 2-hydroxybenzoate

Purchase

Properties

Related Categories Alphabetical Listings, Flavors and Fragrances, G-H More...
biological source   synthetic
InChI Key   DUKPKQFHJQGTGU-UHFFFAOYSA-N
assay   >98%
refractive index   n20/D 1.505(lit.)
bp   290 °C(lit.)
density   1.04 g/mL at 25 °C(lit.)
GMO content   GMO-free
Organoleptic   green
food allergen   no known allergens

Description

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
RTECS 
DH2207000
Flash Point(F) 
251.6 °F
Flash Point(C) 
122 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?