EMAIL THIS PAGE TO A FRIEND
W521205 Aldrich

Cassia oil

redistilled

Synonym: Cinnamomum cassia

Purchase

Properties

Related Categories A-D, Alphabetical Listings, C-D, Flavors and Fragrances, Kosher Certified Products More...
biological source   from Cinnamomum cassia Blume
grade   redistilled
optical activity   [α]20/D −0.5°, neat
refractive index   n20/D 1.614
Organoleptic   woody

Description

Preparation Note

Extraction method: steam distillation

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN3082 - class 9 - PG 3 - DOT NA1993 - Environmentally hazardous substances, liquid, n.o.s. HI: all (not BR)
WGK Germany 
3
Flash Point(F) 
143.6 °F
Flash Point(C) 
62 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Food Safety

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?