EMAIL THIS PAGE TO A FRIEND
W521418 Aldrich

Cedrol

redistilled

Synonym: (+)-Cedrol, (1S,2R,5S,7R,8R)-2,6,6,8-Tetramethyltricyclo[5.3.1.01.5]undecan-8-ol

Purchase

Properties

Related Categories A-D, Alphabetical Listings, C-D, Flavors and Fragrances, Kosher Certified Products More...
biological source   from Juniperus mexicana
  synthetic
grade   Kosher
  redistilled
InChI Key   SVURIXNDRWRAFU-OGMFBOKVSA-N
origin   USA origin
refractive index   n20/D 1.509-1.515
bp   273 °C(lit.)
mp   86-87 °C(lit.)
GMO content   GMO-free
Organoleptic   woody
food allergen   no known allergens

Description

Packaging

1 kg in glass bottle

5 kg in poly drum

Preparation Note

Extraction method: fractional distillation

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation
Protocols & Articles

Articles

GC Analysis of Fragrance Compounds on SUPELCOWAX® 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Fragrance Compounds on SUPELCOWAX® 10
Keywords: Chromatography, Cosmetics, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?