• USA Home
  • W522201 - Cedarwood oil, Texas

EMAIL THIS PAGE TO A FRIEND
W522201 Aldrich

Cedarwood oil, Texas

Purchase

Properties

Related Categories Alphabetical Listings, C-D, Essential Oils, Flavors and Fragrances
biological source   from Juniperus mexicana
origin   USA origin
refractive index   n20/D 1.505(lit.)
Organoleptic   woody

Description

Preparation Note

Extraction method: steam distillation

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?