• USA Home
  • Z100773 - Natural rubber septa
EMAIL THIS PAGE TO A FRIEND
Z100773 Aldrich
Natural rubber septa
white rubber, mixed set (145 pieces with carrying box)
Purchase
Description
General description
Mixed set contains:
25 each of the ones for 5 mm NMR tubes; 7 mm O.D. glass tubing; and 8 or 9 mm O.D. glass tubing
20 each of the ones for 9 or 10 mm O.D. glass tubing; and 10/30 joints and small test tubes
10 each of the ones for 14/20 joints; 19/22 joints; and 24/40 joints
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z554006 Precision Seal® rubber septa, white, mixed set
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?