• USA Home
  • Z101001 - Aldrich® diazomethane-generator apparatus
EMAIL THIS PAGE TO A FRIEND
Z101001 Aldrich
Aldrich® diazomethane-generator apparatus
O-ring joint
Purchase
Description
General description
MNNG is mutagenic and exposure may cause skin sensitivity. While MNNG is more convenient for the generation of small quantities of diazomethane, Diazald is the preferred reagent for large-scale production of diazomethane. However, the following procedure is verified and recommended for use on a small ~0.3 mmol substrate scale. For larger substrate amounts, one of the Diazald distillation apparatuses should be used to generate diazomethane.
Typical Generator Procedure:
To the outside tube of the Aldrich MNNG diazomethane generation apparatus, add 4-methoxybenzoic acid (0.465 g, 0.300 mmol) and ether (3.0 mL). To the inside tube, add Diazald (0.367 g, 1.71 mmol) and carbitol (1.0 mL). Assemble the two parts and immerse the lower part of the outer tube in an ice bath. After equilibrating to the cooling bath temperature, Slowly inject, dropwise through the septum via a syringe, aqueous KOH (37%, ~1.5 mL).
Shake the apparatus gently by hand to ensure mixing of reactants within the inner tube, while being careful not to allow these reactants to spill into the outer tube. The solution in the outer tube may became yellow in color and will persisted, indicating an excess of diazomethane. After 50 min, open the apparatus. Carefully add solid silicic acid (0.151 g) to the inner tube to destroy unreacted diazomethane. Evaporate the yellow ether solution in the outside tube under a gentle stream of nitrogen affording methyl 4-methoxybenoate as a white solid product (0.490 g, 98.4%). Analysis by GC/MS indicates the material to be analytically pure.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z411736 Aldrich® diazomethane-generator with System 45 compatible connection, for the preparation of diazomethane without ether
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?