• USA Home
  • Z101966 - Dual-bulb ball-tube
EMAIL THIS PAGE TO A FRIEND
Z101966 Aldrich
Dual-bulb ball-tube
Joint: ST/NS 24/40, capacity 500 mL
Purchase
Description
General description
ST/NS: joint on one end to connect to oven flask and a hose connector on the other for connection to the motor drive unit.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?