• USA Home
  • Z102369 - Perfektum® one-way compression-nut stopcock
EMAIL THIS PAGE TO A FRIEND
Z102369 Aldrich
Perfektum® one-way compression-nut stopcock
Female Luer to male Luer lock with large bore, not unidirectional
Purchase
Properties
Related Categories Labware, Stopcocks, Syringe connectors, Syringes and Needles
mfr. no.   Cadence Science
  Micro-Mate
Description
General description
Water tested at 80 psi.
Legal Information
Perfektum is a registered trademark of Cadence Science
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?