• USA Home
  • Z103217 - Heating element for flameless heat gun
EMAIL THIS PAGE TO A FRIEND
Z103217 Aldrich
Heating element for flameless heat gun
parameter 300-650 °F temperature (149-343 °C), AC input 120 V, 5 A
Purchase
Properties
Related Categories Heat Guns, Heating, Labware, Manufacturer Browser, Pamran More...
mfr. no.   Pamran
availability   not available in EU
parameter   300-650 °F temperature (149-343 °C)
Description
General description
Includes wire and mica core.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?