• USA Home
  • Z103446 - Chemist’s Companion
EMAIL THIS PAGE TO A FRIEND
Z103446 Aldrich
Chemist’s Companion
provides multiple tables of practical information
  •  ISBN-10 0471315907
  •  ISBN-13 978-047131590-2
Purchase
Properties
Related Categories Books and Software, Labware, Reference Books More...
publication info   A. Gordon and R. Ford, John Wiley & Sons, Ltd, 1973, 560 pp., hard cover
mfr. no.   Wiley
Description
General description
An invaluable resource that should be in every chemistry laboratory. Provides multiple tables of practical information including glassware cleaning solutions, common solvents for crystallization, and solvents and baths for heating and cooling.
Table of Contents
PARTIAL TABLE OF CONTENTS:
Properties of Solvents and Common Liquids
Azeotropic Data
Empirical Boiling Point-Pressure Relationships
Properties of Selected Gases
Acids and Bases Properties of Atoms and Bonds
Properties of the Elements
Table of Isotopes
Selected Bond Lengths
Selected Bond Strengths
Activation Parameters and Kinetics of Selected Reactions
Optical Materials for Spectroscopy and Photochemistry
Photochemistry Light Sources and Equipment
Chromatography
Simple Preparations of Some Dry Gases
Common Solvents for Crystallization
Solvents for Extraction of Aqueous Solutions
Drying Agents
Solvents and Baths for Heating and Cooling
Approved International Units System and General Constants
Useful Conversion Factors
Wavelength-Wavenumber Conversion
Multiples of Element and Group Weights
Subject Index
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?