• USA Home
  • Z104264 - ReDuRan® hand-washing cream

EMAIL THIS PAGE TO A FRIEND
Z104264 Aldrich

ReDuRan® hand-washing cream

removes dyes with powerful cleaning action

Purchase

Properties

Related Categories Cleaners, Hand Cleaners, Labware More...

Description

General description

Removes dyes with powerful cleaning action. Does not contain abrasives or chlorine. Rub onto skin, then wash off.

Legal Information

ReDuRan is a registered trademark of Stockhausen, Inc.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?