EMAIL THIS PAGE TO A FRIEND
Z104507 Aldrich
PTFE beaker
capacity 50 mL
Purchase
Properties
Related Categories Beakers, Labware, PTFE Beakers More...
capacity   50 mL
Description
General description
Chemically resistant except for molten alkali metal.
Temperature range: Subfreezing to 500°F (260°C). Resists adhesion of practically all materials.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?