• USA Home
  • Z104582 - Scienceware® flow monitor
EMAIL THIS PAGE TO A FRIEND
Z104582 Aldrich
Scienceware® flow monitor
Purchase
Properties
Related Categories Bel-Art, Bel-Art Flow Meters, Flowmeters, Indicators, Labware,
mfr. no.   Bel-Art
Description
General description
Colored paddle wheel spins freely with liquid or gas flow (range: 5 to 95 mL/s.) Made of clear polystyrene. Monitor should not be used with corrosive fluids nor at temperatures above 65 °C.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?