• USA Home
  • Z105546 - Polyethylene gloves
EMAIL THIS PAGE TO A FRIEND
Z105546 Aldrich
Polyethylene gloves
size M, wrist length
Purchase
Properties
Related Categories Gloves, Labware, Polyethylene Gloves, Specialist Gloves
material   polyethylene
  clear gloves
size   M
feature   latex free
  powder-free
packaging   pack of 100 gloves
mfr. no.   Sigma-Aldrich
availability   not available in EU
thickness   1.0 mil
Description
General description
Talc-free. Thin gauge poly film allows keen touch sensitivity and short-term protection from moisture and chemicals. Ambidextrous, disposable. Cannot be used for cell culture applications.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?