• USA Home
  • Z105686 - Spinbar® magnetic stir bar
EMAIL THIS PAGE TO A FRIEND
Z105686 Aldrich
Spinbar® magnetic stir bar
PTFE-coated egg-shaped, size 1 5/8 in. × 3/4 in., white
Purchase
Properties
Related Categories 41mm to 60mm Length Oval Stir Bars, Bel-Art, Bel-Art Magnetic Stirring Bars, Bel-Art Spinbar Magnetic Stirring Bars, Labware,
material   PTFE-coated egg-shaped
mfr. no.   Bel-Art
size   1 5/8 in. × 3/4 in.
color   white
Description
General description
Leakproof, seamless, polished, and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.
Legal Information
Spinbar is a registered trademark of SP Scienceware, a division of SP
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?