• USA Home
  • Z106321 - Wheaton connecting adapter
EMAIL THIS PAGE TO A FRIEND
Z106321 Aldrich
Wheaton connecting adapter
To female Clear-Seal® joint, Joint: ST/NS 24/40
Synonym: vacuum adapter, vacuum adapters, wheaton adapters
Purchase
Properties
Related Categories Alcan Packaging, Alcan Packaging Adapters, Glassware, Glassware Adapters, Labware,
Joint   Joint: ST/NS24/40
mfr. no.   Wheaton Scientific
Description
General description
13-425 screw-threaded connector.
Legal Information
Clear-Seal is a registered trademark of Wheaton Industries, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?