EMAIL THIS PAGE TO A FRIEND
Z107190 Aldrich
Base plates
Swivel
Purchase
Description
General description
Fits ½ in. lattice rods. Flat base.
These base plates are of a non-ferrous alloy, two inches in diameter, 16 mm high, with set screw for fastening lattice frame rods. Provided without mounting screws. Plate has four holes for attaching to bench, floor or wall.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?