EMAIL THIS PAGE TO A FRIEND
Z108030 Aldrich
Rubber dam
pkg of 1 m
Purchase
Properties
Related Categories Buchner Funnels, Funnels, Labware More...
packaging   pkg of 1 m
Description
General description
For use with Buchner funnels. A 42 in. wide latex sheet that aids in the thorough removal of liquid from a filter cake in a Buchner funnel (after vacuum filtration).
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?