• USA Home
  • Z108200 - Compendium Of Organic Synthetic Methods
EMAIL THIS PAGE TO A FRIEND
Z108200 Aldrich
Compendium Of Organic Synthetic Methods
Vol. IV
  •  ISBN-10 0471049239
  •  ISBN-13 978-047104923-4
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   M. Smith, 1980, 520 pp., hard cover
mfr. no.   Wiley
Description
General description
Each updated volume presents the latest synthetic methods for the preparation of mono- and difunctional compounds.
Table of Contents
Abbreviations.
Index, Monofunctional Compounds.
Index, Difunctional Compounds.
Introduction.
Preparation of Acetylenes.
Preparation of Carboxylic Acids, Acid Halides, and Anhydrides.
Preparation of Alcohols and Phenols.
Preparation of Aldehydes.
Preparation of Alkyls, Methylenes, and Aryls.
Preparation of Amides.
Preparation of Amines.
Preparation of Esters.
Preparation of Ethers and Epoxides.
Preparation of Halides and Sulfonates.
Preparation of Hydrides.
Preparation of Ketones.
Preparation of Nitriles.
Preparation of Olefins.
Preparation of Difunctional Compounds.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?