• USA Home
  • Z108391 - Window for Aldrich® adjustable cell holder
EMAIL THIS PAGE TO A FRIEND
Z108391 Aldrich
Window for Aldrich® adjustable cell holder
NaCl, Polished
Purchase
Properties
Related Categories Crystal Windows, Labware, Spectroscopy Supplies, Spectroscopy, IR
material   NaCl
  rectangular
W × L × H   11 mm × 30 mm × 7 mm
Description
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?