• USA Home
  • Z108448 - NORTH Viton® fluoroelastomer gloves
EMAIL THIS PAGE TO A FRIEND
Z108448 Aldrich
NORTH Viton® fluoroelastomer gloves
size 10, standard, L × thickness 11 in. × 10 mil
Purchase
Properties
Related Categories Fluorocarbon & Other Fluorocompound Heavyweight Gloves, Gloves, Heavyweight Gloves, Labware, Manufacturer Browser,
size   10
packaging   pack of 1 pair
mfr. no.   North
L × thickness   11 in. × 10 mil
Description
General description
Outstanding chemical resistance and a high degree of impermeability to many hazardous solvents. Virtually unsurpassed by any other glove type. Designed especially for handling aromatic and chlorinated solvents.
Legal Information
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?