• USA Home
  • Z108766 - Annual Reports In Organic Synthesis
EMAIL THIS PAGE TO A FRIEND
Z108766 Aldrich
Annual Reports In Organic Synthesis
1979
  •  ISBN-10 0120408104
  •  ISBN-13 978-012040810-8
Purchase
Properties
Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   J. McMurray, ed., 1980, soft cover
mfr. no.   Elsevier
Description
General description
Abstracts synthetic reactions from the major chemistry journals of the past year and includes all reactions and methods that are new and reasonably general. The reactions are presented in a convenient pictorial format designed for rapid visual retrieval of information.
Table of Contents
Carbon-Carbon Bond Forming Reactions.
Oxidations.
Reductions.
Synthesis of Heterocycles.
Protecting Groups.
Useful Synthetic Preparations.
Reviews.
Author Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?