EMAIL THIS PAGE TO A FRIEND
Z108863 Aldrich
Flameless heat gun
AC input 230 V, parameter 500-650 °F temp. range (260-343 °C)
Purchase
Properties
Related Categories Heat Guns, Heating, Labware More...
mfr. no.   Pamran
parameter   500-650 °F temp. range (260-343 °C)
output   2.5 A
Description
General description
Adjustable shutter allows temperature regulation. High-velocity, self-lubricated motor is rubber-mounted for quiet, vibration-free handling. Triple-plated chrome steel barrel and housing. Double-insulated barrel has grilled nozzle guard for safety.
Recessed, three-way switch prevents accidental turn-on. Includes stand, storage chest, and 8 ft, 3-wire cord.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?