• USA Home
  • Z109681 - Hamilton® 800 Series MULTI-PAK
EMAIL THIS PAGE TO A FRIEND
Z109681 Aldrich
Hamilton® 800 Series MULTI-PAK
800RNW, volume 10-250 μL, needle size 25s ga (blunt tip), needle L 51 mm (2 in.), pkg of 5 syringes
Purchase
Properties
Related Categories 800 Series Syringes, Analytical/Chromatography, GC Manual Syringes, Hamilton Microliter 800 Series Syringe, Hamilton Microliter Syringes, 800 and 900 Series,
needle size   25s ga (blunt tip)
packaging   pkg of 5 syringes
mfr. no.   Hamilton, 84891
needle L   51 mm (2 in.)
volume   10-250 μL
Description
General description
Designed for Waters® U6K Injection Valve.
Includes one each of 10, 25, 50, 100, & 250μL volume syringes:
• Contains one handle and a selection of syringe plunger/barrel combinations
• Removable needles (RNW), includes nine needles
• Reinforced plunger - plunger bending is eliminated due to the reinforced plunger stem.
Items included in the 800 series Multi-Pak (Hamilton part number 84890) include:
Hamilton part number Description
• 32135 Handle for barrel/plunger
• 32129 801RN barrel/plunger, 10 μL
• 32117 802RN barrel/plunger, 25 μL
• 32120 805RN barrel/plunger, 50 μL
• 32123 810RN barrel/plunger, 100 μL
• 32126 825RN barrel/plunger, 250 μL
• 30902 RN nut
• 7758-02 Removable needles (26s/2"/2), 6/pk for 10-100 μL syringes
• 7758-03 Removable needles (22s/2"/2), 6/pk for 10-100 μL syringes
• 7779-03 Removable needles (22s/2"/2), 6/pk for 250 μL syringes
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Waters is a registered trademark of Waters
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?