• USA Home
  • Z110051 - Thermco® PTFE-coated thermometer
EMAIL THIS PAGE TO A FRIEND
Z110051 Aldrich
Thermco® PTFE-coated thermometer
0-230 °F temperature
Purchase
Properties
Related Categories Labware, Temperature, Thermometers More...
feature   graduations 2 °F
mfr. no.   Thermco Products, Inc.
measuring range   0-230 °F temperature
L   300 mm
Description
General description
76 mm immersion thermometer within PTFE jacket. Impervious to acids and corrosive chemicals, shatter-resistant, and shock-absorbent. Contains Hg fill.
Legal Information
Thermco is a registered trademark of Thermco Products, Inc.
Price and Availability
Safety & Documentation
Safety Information
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?