• USA Home
  • Z110086 - Thermco® PTFE-coated thermometer
EMAIL THIS PAGE TO A FRIEND
Z110086 Aldrich
Thermco® PTFE-coated thermometer
20-400 °F temperature
Purchase
Properties
Related Categories Labware, Temperature, Thermometers More...
feature   graduations 2 °F
mfr. no.   Thermco Products, Inc.
measuring range   20-400 °F temperature
L   300 mm
Description
General description
76 mm immersion thermometer within PTFE jacket. Impervious to acids and corrosive chemicals, shatter-resistant, and shock-absorbent. Contains Hg fill.
Legal Information
Thermco is a registered trademark of Thermco Products, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?