• USA Home
  • Z110094 - Maximum-minimum thermometer
EMAIL THIS PAGE TO A FRIEND
Z110094 Aldrich
Maximum-minimum thermometer
for indoor or outdoor use, parameter -40-120 °F temp. limit
Purchase
Properties
Related Categories Labware, Temperature, Thermometers More...
parameter   -40-120 °F temp. limit
Description
General description
Registers maximum and minimum temperatures in both °C (range: −40 to 50° with 1° divisions) and °F (range: −40 to 120° with 2° divisions). Hg filled bulbs sheltered in casing for indoor or outdoor use. Can be reset by depressing button.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?