• USA Home
  • Z110345 - Thermco® ground-glass joint thermometer
EMAIL THIS PAGE TO A FRIEND
Z110345 Aldrich
Thermco® ground-glass joint thermometer
−10-250 °C temperature, immersion level, 150 mm
Purchase
Properties
Related Categories Labware, Temperature, Thermometers More...
feature   immersion level 150 mm
mfr. no.   Thermco Products, Inc.
measuring range   −10-250 °C temperature
Description
General description
ST/NS: 10/30 ground-glass joint. All have 1° divisions. Contains Hg fill.
Legal Information
Thermco is a registered trademark of Thermco Products, Inc.
Price and Availability
Safety & Documentation
Safety Information
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?