• USA Home
  • Z110396 - Thermco® adjustable-bushing thermometer
EMAIL THIS PAGE TO A FRIEND
Z110396 Aldrich
Thermco® adjustable-bushing thermometer
-10-360 °C temperature, immersion level, ≤ 6 in.
Purchase
Properties
Related Categories Labware, Temperature, Thermometers More...
feature   immersion level≤ 6 in.
mfr. no.   Thermco Products, Inc.
measuring range   -10-360 °C temperature
Description
General description
Equipped with an adjustable bushing to provide a variety of immersion lengths. Bushing consists of a glass sleeve with a ST/NS: 10/18 ground-glass inner joint. A rubber cap holds the thermometer at the desired immersion length. All have 1° divisions. Contains Hg fill.
Legal Information
Thermco is a registered trademark of Thermco Products, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?