• USA Home
  • Z110426 - Bushing for adjustable-bushing thermometer
EMAIL THIS PAGE TO A FRIEND
Z110426 Aldrich
Bushing for adjustable-bushing thermometer
fits either 6" or 12" immersion length
Purchase
Properties
Related Categories Labware, Temperature, Thermometers More...
packaging   pkg of 1 ea
Description
General description
For either immersion length (replacement bushing only)
• Thermometers have 10/18 Bushing
• Bushing slides along stem allowing many immersion lengths
• 6” immersion for flask sizes: 250 to 3000 mL
• 12” immersion for flask sizes: 5,000 to 22,000 mL
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?