• USA Home
  • Z110930 - Thermolyne® Nuova stirrer
EMAIL THIS PAGE TO A FRIEND
Z110930 Aldrich
Thermolyne® Nuova stirrer
AC/DC input 120 V, 0.3 A, output25 W
Synonym: magnetic stirrers, stirrer, stirrers
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Stirrers, Labware, Magnetic Stirrers, Manufacturer Browser,
mfr. no.   Barnstead International S18525
availability   not available in EU
  not available in EU
speed   60-1000 rpm
L × W × H   11 in. × 7 1/4 in. × 3 1/2 in., overall
output   25 W
Description
General description
Solid-state speed control, chemically resistant porcelain-coated SS top plate (7 × 7 in.). Direct drive motor gives smooth stirring at low speeds and thorough churning action at high speeds. Includes 1 in. diam. star head and 3/8 diam. × 2 in. stirring bar.
Legal Information
Nuova is a trademark of Barnstead/Thermolyne Corp.
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?