• USA Home
  • Z110949 - Thermolyne® Nuova stirrer
EMAIL THIS PAGE TO A FRIEND
Z110949 Aldrich
Thermolyne® Nuova stirrer
AC/DC input 240 V AC, 0.1 A, output17 W
Synonym: magnetic stirrers, stirrer, stirrers
Purchase
Properties
Related Categories Labware, Magnetic Stirrers, Stirrers More...
mfr. no.   Barnstead International S1852026
availability   not available in EU
speed   60-1000 rpm
L × W × H   11 in. × 7 1/4 in. × 3 1/2 in., overall
output   17 W
Description
General description
Solid-state speed control, chemically resistant porcelain-coated SS top plate (7 × 7 in.). Direct drive motor gives smooth stirring at low speeds and thorough churning action at high speeds. Includes 1 in. diam. star head and 3/8 diam. × 2 in. stirring bar.
Legal Information
Nuova is a trademark of Barnstead/Thermolyne Corp.
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?