• USA Home
  • Z110957 - Thermolyne® Nuova stirring hotplate

EMAIL THIS PAGE TO A FRIEND
Z110957 Aldrich

Thermolyne® Nuova stirring hotplate

AC input 120 V

Purchase

Properties

Related Categories Barnstead Thermolyne, Barnstead Thermolyne Heating Equipment, Barnstead Thermolyne Hot Plates, Heating, Hotplate Stirrers,
sterility   non-sterile
mfr. no.   Barnstead International SP18425
availability   not available in EU
output   858 watts, 7.3 A
color   none
matrix active group   Non-treated surface treatment

Description

General description

Has same features as NUOVA hot plate plus direct drive motor for smooth stirring at low speeds and thorough churning at higher speeds. Stirring speed range: 100 to 1,000 rpm. UL listed.

Legal Information

Nuova is a trademark of Barnstead/Thermolyne Corp.

Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?