• USA Home
  • Z111007 - Thermolyne® hotplate oven

EMAIL THIS PAGE TO A FRIEND
Z111007 Aldrich

Thermolyne® hotplate oven

Purchase

Properties

Related Categories Barnstead Thermolyne, Barnstead Thermolyne Heating Equipment, Barnstead Thermolyne Hot Plates, Heating, Hotplates,
AC input   115 V
sterility   non-sterile
mfr. no.   Barnstead International OV10600
L × W × H   8 1/2 in. × 8 3/4 in. × 7 3/8 in.
chamber L × W × H   6 in. × 6 in. × 6 in.
color   none
matrix active group   Non-treated surface treatment

Description

General description

Converts Type 1900 hot plate into a small utility oven. Double-wall construction, steel case, and aluminum interior with air space provide optimum insulation. Heavy aluminum chamber provides excellent heat distribution; max. operating temp.: 300 °C. Thermometer port on top. Note: Hot plate NOT included.

Legal Information

Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?