• USA Home
  • Z111015 - Thermolyne® micro hotplate
EMAIL THIS PAGE TO A FRIEND
Z111015 Aldrich
Thermolyne® micro hotplate
AC/DC input 120 V
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Heating Equipment, Barnstead Thermolyne Hot Plates, Heating, Hotplates,
mfr. no.   Barnstead International HP2305B
availability   not available in EU
output   325 watts, 2.7 A
Description
General description
Contains a demand-type thermostat offering a wide selection of temperature settings from ambient to 371±5 °C. Automatically adjusts for ambient temperature and line-voltage fluctuations. Heating elements bring cast aluminum top plate (41/2 in. diam.) to 260 °C in 41/2 min and to the max. 371 °C in 71/2 min. Perforated SS case allows air circulation around components. 33/4 in. H, 13/4 lb. Includes 3-wire cord, plug, and dovetail socket.
Legal Information
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?