• USA Home
  • Z111023 - Thermolyne® micro hotplate

EMAIL THIS PAGE TO A FRIEND
Z111023 Aldrich

Thermolyne® micro hotplate

AC input 240 V

Purchase

Properties

Related Categories Barnstead Thermolyne, Barnstead Thermolyne Heating Equipment, Barnstead Thermolyne Hot Plates, Heating, Hotplates,
sterility   non-sterile
mfr. no.   Barnstead International HP2310B
output   305 watts, 1.3 A
color   none
matrix active group   Non-treated surface treatment

Description

General description

Contains a demand-type thermostat offering a wide selection of temperature settings from ambient to 371±5 °C. Automatically adjusts for ambient temperature and line-voltage fluctuations. Heating elements bring cast aluminum top plate (41/2 in. diam.) to 260 °C in 41/2 min and to the max. 371 °C in 71/2 min. Perforated SS case allows air circulation around components. 33/4 in. H, 13/4 lb. Includes 3-wire cord, plug, and dovetail socket.

Legal Information

Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?