• USA Home
  • Z111155 - Thermolyne® midget stir plate

EMAIL THIS PAGE TO A FRIEND
Z111155 Aldrich

Thermolyne® midget stir plate

max. temp. of 260°C in 10 min

Purchase

Properties

Related Categories Barnstead Thermolyne, Barnstead Thermolyne Heating Equipment, Barnstead Thermolyne Hot Plates, Labware, Magnetic Stirrers,
AC input   120 V
sterility   non-sterile
mfr. no.   Barnstead International SP10105B
availability   not available in EU
L × W × H   3 5/8 in. × 4 1/4 in. × 4 1/4 in.
plate size   4 1/2 in. × 4 1/2 in.
output   300 watts, 2.5 A
color   none
matrix active group   Non-treated surface treatment

Description

General description

Similar to Type 1000 stir plate, but without solid-state speed control. Reaches max. temp. of 260°C in 10 min. Includes 1 in. diam., star-head PTFE-coated stirring bar and dovetail bracket. UL listed.

Legal Information

Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?