• USA Home
  • Z111198 - Thermolyne® type 7200 stirrer
EMAIL THIS PAGE TO A FRIEND
Z111198 Aldrich
Thermolyne® type 7200 stirrer
AC/DC input 240 V AC, 0.18 A, output42 W
Purchase
Properties
Related Categories Labware, Magnetic Stirrers, Stirrers More...
mfr. no.   Barnstead International S7220
speed   60-1200 rpm
stirrer H   4 1/2 in.
output   42 W
Description
General description
Solid-state speed control, aluminum top plate 7 × 7 in. Has perforated SS case for cool, extended stirring. Capable of stirring large volumes and viscous solutions. Includes 1 in. diam. star head and 3/8 diam. × 2 in. stirring bar.
Legal Information
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?