• USA Home
  • Z111236 - Thermolyne® Stir-Mate stirrer
EMAIL THIS PAGE TO A FRIEND
Z111236 Aldrich
Thermolyne® Stir-Mate stirrer
output18 W
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Stirrers, Labware, Magnetic Stirrers, Manufacturer Browser,
AC input   120 V, 0.15 A
mfr. no.   Barnstead International S7805
speed   0-1200 rpm
diam. × H   4 1/2 in. × 3 in.
weight   1.5 lb
output   18 W
Description
General description
On/off switch preserves speed setting on stepless speed control dial. SS case, small and light. Ideal for confined spaces. Will stir through most heating mantles. Includes 3/8 diam. × 2 in. stirring bar.
Legal Information
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z403644 IKA® Color Squid IKAMAG magnetic plate stirrers, small magnetic stirrer, Number One top plate design, 1/cs
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?