• USA Home
  • Z111244 - Thermolyne® Test Tube Maxi-Mix
EMAIL THIS PAGE TO A FRIEND
Z111244 Aldrich
Thermolyne® Test Tube Maxi-Mix
AC input 120 V
Purchase
Properties
Related Categories Barnstead Thermolyne, Barnstead Thermolyne Shakers, Labware, Manufacturer Browser, Shakers & Vortexers,
mfr. no.   Barnstead International M16715
Description
General description
Accommodates 4 or more test tubes or small flasks. Mixing action is varied by changing pressure of test tubes against foam-rubber top. Has top mounted pushbutton on/off switch, counterbalance, and suction-cup feet. Compact, 41/8 in. H with 4 in. diam. top.
Legal Information
Maxi-Mix is a trademark of Barnstead/Thermolyne Corp.
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z645001 IKA® MS 3 basic shakers, universal small shaker, 115 V
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?