• USA Home
  • Z111465 - Polymer Syntheses, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
Z111465 Aldrich
Polymer Syntheses, 2nd ed.
Volume 3
  •  ISBN-10 0126185131
  •  ISBN-13 978-012618513-3
Purchase
Properties
Related Categories Books and Software, Labware, Polymer Chemistry Books More...
publication info   , 1996, case bound
mfr. no.   Elsevier
Description
General description
Provides detailed laboratory instructions for the synthesis of polymers.
This volume, which completes the Second Edition of this widely respected three-volume set, continues the tradition of providing detailed and concise laboratory instructions for the preparation of organic and inorganic polymers. Updates are provided on the preparation of various types of polymers including olefin-sulfur dioxide copolymers, polythioesters, sulfide polymers, polyisocyanates, polyoxyalkylhydroxy compounds, polyvinyl carbazole, polyvinyl acetate, polyallyl esters, and polyvinyl flouride. The procedures were chosen on the basis of safety and to be representative of preparation methods for a general class of polymers. Fundamental and practical information on polymer synthesis is combined with extensive references and reviews of the patent literature.
Table of Contents
Preface.
Olefin-Sulfur Dioxide Copolymers.
Polythioesters.
Sulfide Polymers.
Polymerization Reactions of Mono- and Diisocyanates.
Polyoxyalkylation of Hydroxy Compounds.
Polymerization Reactions of N-Vinyl Carbazole and Related Monomers.
Polymerization of Vinyl Acetate and Other Vinyl Esters.
Polymerization of Allyl Esters.
Polymerization of Vinyl Floride.
Miscellaneous Polymer Preparations.
Author Index.
Subject Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?